siRNA Details
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 1 | 480 | ||
GenBank mRNA | AK039944.1 | - | 1 | 484 | ||
AK050306.1 | - | 1 | 481 | |||
AK053715.1 | - | 1 | 480 | |||
AK084697.1 | - | 1 | 484 | |||
AK086395.1 | - | 1 | 483 | |||
AK173211.1 | - | 1 | 459 | |||
BC150785.1 | - | 1 | 474 | 9087696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
