siRNA Details
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018729.2 | NP_061199.2 | 9 | 2175 | ||
GenBank mRNA | AK033178.1 | - | 4 | 1158 | ||
AK155548.1 | - | 9 | 2122 | |||
AK172371.1 | - | 1 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
