siRNA Details
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146003.2 | NP_666115.2 | 2, 3 | 756 | ||
GenBank mRNA | AK053904.1 | - | 2, 3 | 756 | ||
AK083377.1 | - | 2, 3 | 345 | |||
BC061480.1 | - | 2, 3 | 580 | 6509771 | ||
BC151128.1 | - | 2, 3 | 166 | 9088041 | ||
BC172171.1 | - | 2, 3 | 166 | 9054841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
