siRNA Details
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 3 | 307 | ||
GenBank mRNA | AF435012.1 | - | 3 | 291 | ||
AK052521.1 | - | 3 | 276 | |||
AK078542.1 | - | 3 | 308 | |||
AK084394.1 | - | 4 | 487 | |||
AK084811.1 | - | 3 | 305 | |||
AK145252.1 | - | 3 | 308 |

More Information
Other Species Targeted

Gene Ontology Categories:
