siRNA Details
Gene Symbol: Fbxl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084174.1 | - | 1 | 926 | ||
AK084297.1 | - | 1 | 380 | |||
BC050864.1 | - | 2 | 1433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
