siRNA Details
Gene Symbol: Igf2bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009951.4 | NP_034081.1 | 15 | 7070 | ||
GenBank mRNA | AK050850.1 | - | 1 | 1835 | ||
AK083554.1 | - | 1 | 1830 | |||
BC052759.1 | - | 1 | 786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
