siRNA Details
Gene Symbol: Zfp608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175751.4 | NP_786927.2 | 7 | 4692 | ||
GenBank mRNA | AK076178.1 | - | 3 | 1568 | ||
AK077030.1 | - | 6 | 3419 | |||
AK129321.1 | - | 2 | 369 | |||
BC059268.1 | - | 6 | 4358 | |||
BC092219.1 | - | 7 | 4425 | 6830664 | ||
BC151073.1 | - | 6 | 4211 | 9087986 | ||
BC151075.1 | - | 6 | 4211 | 9087988 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107378.
Gene Ontology Categories:
