siRNA Details
Gene Symbol: Ttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011652.3 | NP_035782.3 | 9 | 1689 | ||
NM_028004.2 | NP_082280.2 | 9 | 1689 | |||
GenBank mRNA | AK052859.1 | - | 9 | 1689 | ||
AK084709.1 | - | 9 | 1689 | |||
AK084718.1 | - | 9 | 1685 | |||
AK085832.1 | - | 9 | 1689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
