siRNA Details
Gene Symbol: DNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130823.2 | NP_001124295.1 | 1, 2 | 321 | ||
NM_001318730.1 | NP_001305659.1 | 1, 2 | 240 | |||
NM_001318731.1 | NP_001305660.1 | 1, 2 | 240 | |||
NM_001379.3 | NP_001370.1 | 1, 2 | 321 | |||
GenBank mRNA | AB209413.2 | - | 1, 2 | 47 | ||
BC092517.1 | - | 1, 2 | 120 | 3077831 | ||
BC126227.1 | - | 1, 2 | 265 | 8991943 | ||
BC144093.1 | - | 1, 2 | 265 | 9052613 | ||
X63692.1 | - | 1, 2 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
