siRNA Details
Gene Symbol: Ccdc88a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176841.4 | NP_789811.2 | 2 | 275 | ||
GenBank mRNA | AB087827.1 | - | 2 | 99 | ||
AK051140.1 | - | 2 | 673 |

More Information
Other Species Targeted

Gene Ontology Categories:
