siRNA Details
Gene Symbol: Ice2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145618.3 | NP_663593.2 | 4 | 390 | ||
GenBank mRNA | AK041772.1 | - | 4 | 447 | ||
AK046121.1 | - | 4 | 452 | |||
AK134186.1 | - | 4 | 435 | |||
AY244558.1 | - | 4 | 419 | |||
BC068183.1 | - | 4 | 390 | 30639735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
