siRNA Details
Gene Symbol: Auts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177047.3 | NP_796021.2 | 1 | 696 | ||
GenBank mRNA | AK052418.1 | - | 1 | 758 | ||
AK077988.1 | - | 1 | 1153 | |||
AK085946.1 | - | 1 | 385 | |||
AK147478.1 | - | 1695 | ||||
BC096021.1 | - | 1 | 695 | |||
BC151031.1 | - | 1 | 710 | |||
BC151047.1 | - | 1 | 709 |

More Information
Other Species Targeted

Gene Ontology Categories:
