siRNA Details
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 17 | 4077 | ||
GenBank mRNA | AK083984.1 | - | 1 | 170 | ||
AK088853.1 | - | 1 | 781 | |||
AK122406.1 | - | 16 | 5022 | |||
AW911453.1 | - | 1 | 408 | |||
BC051527.1 | - | 1 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
