siRNA Details
Gene Symbol: 9430076C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034642.1 | - | 149 | |||
AK035031.1 | - | 3 | 200 | |||
AK051275.1 | - | 3 | 174 | |||
AK166503.1 | - | 3 | 186 |
Gene Symbol: Creb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084024.1 | - | 80 | |||
AK170766.1 | - | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
