siRNA Details
Gene Symbol: Ccdc88a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176841.4 | NP_789811.2 | 2, 3 | 334 | ||
GenBank mRNA | AB087827.1 | - | 2, 3 | 158 | ||
AK051140.1 | - | 2, 3 | 732 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770470, FQ230491.
Gene Ontology Categories:
