siRNA Details
Gene Symbol: Myof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099634.1 | NP_001093104.1 | 6 | 703 | ||
GenBank mRNA | AK031666.1 | - | 6 | 526 | ||
AK086107.1 | - | 6 | 485 | |||
AK087176.1 | - | 6 | 526 | |||
AK087302.1 | - | 6 | 504 | |||
AK173131.1 | - | 6 | 484 | |||
BC150790.1 | - | 6 | 477 | 9087701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
