siRNA Details
Gene Symbol: Trappc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029491.2 | NP_083767.2 | 2 | 347 | ||
NM_177038.3 | NP_796012.2 | 2 | 347 | |||
GenBank mRNA | AK083747.1 | - | 2 | 347 |

More Information
Other Species Targeted

Gene Ontology Categories:
