siRNA Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081288.1 | NP_001074757.1 | 5 | 657 | ||
GenBank mRNA | AK028828.1 | - | 5 | 404 | ||
AK147903.1 | - | 5 | 658 | |||
BC147638.1 | - | 5 | 608 | 9056628 | ||
BC151079.1 | - | 5 | 608 | 9087992 |

More Information
Other Species Targeted

Gene Ontology Categories:
