siRNA Details
Gene Symbol: Auts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177047.3 | NP_796021.2 | 1 | 783 | ||
GenBank mRNA | AK052418.1 | - | 1 | 845 | ||
AK077988.1 | - | 1 | 1240 | |||
AK085946.1 | - | 1 | 472 | |||
AK147478.1 | - | 1782 | ||||
BC096021.1 | - | 1 | 782 | |||
BC151031.1 | - | 1 | 797 | |||
BC151047.1 | - | 1 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
