siRNA Details
Gene Symbol: A630001G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177055.3 | NP_796029.2 | 3 | 374 | ||
GenBank mRNA | AA289536.1 | - | 3 | 373 | ||
AK041324.1 | - | 3 | 345 | |||
AK041675.1 | - | 3 | 236 | |||
AK139572.1 | - | 493 | ||||
BB871611.1 | - | 3 | 342 | |||
BC052931.1 | - | 3 | 342 | 1066663 | ||
BG276509.1 | - | 2 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
