siRNA Details
Gene Symbol: Fbxw16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177070.3 | NP_796044.2 | 7 | 933 | ||
GenBank mRNA | AK078661.1 | - | 7 | 935 | ||
BC100319.1 | - | 7 | 917 | 30940137 | ||
DV662133.1 | - | 2 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
