siRNA Details
Gene Symbol: Creb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084024.1 | - | 250 | |||
AK170766.1 | - | 324 |
Gene Symbol: 9430076C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034642.1 | - | 319 | |||
AK035031.1 | - | 4 | 370 | |||
AK051275.1 | - | 4 | 344 | |||
AK166503.1 | - | 4 | 356 |

More Information
Other Species Targeted

