siRNA Details
Gene Symbol: MAST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319245.1 | NP_001306174.1 | 2 | 503 | ||
NM_001324320.1 | NP_001311249.1 | 2 | 509 | |||
NM_015112.2 | NP_055927.2 | 2 | 503 | |||
XM_011541059.2 | XP_011539361.1 | 2 | 521 | |||
XM_011541061.2 | XP_011539363.1 | 2 | 521 | |||
XM_011541062.2 | XP_011539364.1 | 2 | 521 | |||
XM_011541063.1 | XP_011539365.1 | 2 | 85 | |||
XM_011541064.2 | XP_011539366.1 | 2 | 82 | |||
XM_011541065.1 | XP_011539367.1 | 2 | 184 | |||
XM_011541069.2 | XP_011539371.1 | 2 | 521 | |||
GenBank mRNA | AB047005.1 | - | 2 | 503 | ||
BC065499.1 | - | 2 | 477 | 5499830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
