siRNA Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081288.1 | NP_001074757.1 | 9 | 1344 | ||
GenBank mRNA | AK028828.1 | - | 9 | 1091 | ||
AK147903.1 | - | 9 | 1346 | |||
BC147638.1 | - | 9 | 1295 | 9056628 | ||
BC151079.1 | - | 9 | 1295 | 9087992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
