siRNA Details
Gene Symbol: Ercc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081221.1 | NP_001074690.1 | 21 | 5736 | ||
GenBank mRNA | AK040645.1 | - | 1 | 1035 | ||
AK048416.1 | - | 4 | 2195 | |||
AK139396.1 | - | 1 | 466 | |||
BC132447.1 | - | 17 | 4675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
