siRNA Details
Gene Symbol: A630001G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177055.3 | NP_796029.2 | 6 | 961 | ||
GenBank mRNA | AK041324.1 | - | 6 | 1139 | ||
AK041675.1 | - | 6 | 823 | |||
AK139572.1 | - | 1080 | ||||
BC052931.1 | - | 6 | 928 | 1066663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
