siRNA Details
Gene Symbol: Fbxw19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177703.3 | NP_808371.1 | 8 | 1077 | ||
GenBank mRNA | AK087808.1 | - | 8 | 1078 |
Gene Symbol: Fbxw21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177069.3 | NP_796043.1 | 8 | 1103 | ||
GenBank mRNA | AK087710.1 | - | 8 | 1104 | ||
AK135949.1 | - | 8 | 1104 | |||
AK136136.1 | - | 8 | 1100 | |||
BC099397.1 | - | 8 | 1085 | 30936381 |
Gene Symbol: Fbxw25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143305.1 | - | 8 | 1100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
