siRNA Details
Gene Symbol: L3mbtl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177278.5 | NP_796252.2 | 21 | 3805 | ||
GenBank mRNA | AK042907.1 | - | 3 | 1612 | ||
AK053993.1 | - | 2 | 2025 | |||
AK139210.1 | - | 3 | 1613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
