siRNA Details
Gene Symbol: Cntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177129.5 | NP_796103.2 | 23 | 8174 | ||
GenBank mRNA | AK083808.1 | - | 1 | 2457 | ||
AK084013.1 | - | 2, 3 | 1481 | |||
BC079602.1 | - | 1 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
