siRNA Details
Gene Symbol: Vps13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177151.3 | NP_796125.2 | 2, 3 | 415 | ||
GenBank mRNA | AK053274.1 | - | 2, 3 | 415 | ||
BC158052.1 | - | 2, 3 | 395 |

More Information
Other Species Targeted

Gene Ontology Categories:
