siRNA Details
Gene Symbol: B330016D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043314.1 | - | 1 | 379 | ||
AK046111.1 | - | 1 | 1008 | |||
AK046532.1 | - | 1 | 1004 | |||
AK080800.1 | - | 1 | 1015 | |||
AK080864.1 | - | 1 | 984 | |||
AK082239.1 | - | 1 | 386 | |||
AK142965.1 | - | 1 | 1003 | |||
AK148126.1 | - | 1 | 356 |
