siRNA Details
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081179.1 | NP_001074648.1 | 6 | 781 | ||
GenBank mRNA | AK053037.1 | - | 6 | 765 | ||
AK079542.1 | - | 6 | 726 | |||
AK146166.1 | - | 6 | 762 | |||
AK158478.1 | - | 6 | 766 | |||
AK220390.1 | - | 3 | 314 | |||
BC086652.1 | - | 5 | 717 | 6856641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
