siRNA Details
Gene Symbol: Smg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031814.1 | NP_001026984.1 | 2 | 558 | ||
GenBank mRNA | AK041264.1 | - | 2 | 337 | ||
AK047829.1 | - | 2 | 539 | |||
AK049391.1 | - | 2 | 558 | |||
AK157798.1 | - | 2 | 545 | |||
AK220328.1 | - | 2 | 270 |

More Information
Other Species Targeted

Gene Ontology Categories:
