siRNA Details
Gene Symbol: Gm3286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122678.2 | NP_001116150.1 | 2, 3 | 419 | ||
NM_001167793.1 | NP_001161265.1 | 2, 3 | 459 | |||
GenBank mRNA | AK145737.1 | - | 4, 5 | 715 | ||
CF157544.1 | - | 405 | ||||
CN723928.1 | - | 443 | ||||
DV653222.1 | - | 127 |
Gene Symbol: D5Ertd577e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177187.4 | NP_796161.3 | 2, 3 | 421 | ||
GenBank mRNA | AK054417.1 | - | 2, 3 | 421 | ||
BC053383.1 | - | 1, 2 | 330 | |||
BC094905.1 | - | 1, 2 | 318 | 30047197 | ||
BC099941.1 | - | 2, 3 | 405 | 30031929 | ||
BC145804.1 | - | 2, 3 | 367 | 40131107 |
Gene Symbol: E330014E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122668.1 | NP_001116140.1 | 2, 3 | 541 | ||
GenBank mRNA | AK087741.1 | - | 2, 3 | 542 |
Gene Symbol: Gm16367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031622.2 | NP_001026793.2 | 2, 3 | 446 | ||
GenBank mRNA | AK145705.1 | - | 2, 3 | 459 | ||
BC099967.1 | - | 2, 3 | 446 | 30027969 |
Gene Symbol: Gm16513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200055.1 | NP_001186984.1 | 457 |
Gene Symbol: Gm3106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061196.1 | - | 551 |
Gene Symbol: Gm3139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243937.1 | NP_001230866.1 | 2, 3 | 477 | ||
NM_001243938.1 | NP_001230867.1 | 2, 3 | 437 | |||
GenBank mRNA | CN708470.1 | - | 2, 3 | 462 | ||
CN715963.1 | - | 2, 3 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
