siRNA Details
Gene Symbol: Sycp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177191.3 | NP_796165.2 | 31 | 3045 | ||
GenBank mRNA | AK029735.1 | - | 4 | 356 | ||
AK036624.1 | - | 9 | 893 | |||
DQ103262.1 | - | 31 | 3045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
