siRNA Details
Gene Symbol: Tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077418.1 | - | 2 | 266 | ||
AK080378.1 | - | 2 | 258 | |||
AK169483.1 | - | 2 | 258 | |||
BC057996.1 | - | 2 | 168 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008768790, XM_008768789.
