siRNA Details
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011505.2 | NP_035635.1 | 18 | 5215 | ||
GenBank mRNA | AK031650.1 | - | 1 | 2553 | ||
AK081165.1 | - | 1 | 1600 | |||
BC090673.1 | - | 1 | 1780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
