siRNA Details
Gene Symbol: Spef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041992.1 | - | 3 | 397 | ||
AK048961.1 | - | 3 | 488 | |||
BC132098.1 | - | 3 | 425 | 40130375 | ||
BC132288.1 | - | 3 | 425 | 40130565 | ||
HQ856050.1 | - | 3 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
