siRNA Details
Gene Symbol: Vps13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177151.3 | NP_796125.2 | 3, 4 | 559 | ||
GenBank mRNA | AK049111.1 | - | 4, 5 | 679 | ||
AK053274.1 | - | 3, 4 | 559 | |||
BC158052.1 | - | 3, 4 | 539 |

More Information
Other Species Targeted

Gene Ontology Categories:
