siRNA Details
Gene Symbol: PASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252124.1 | NP_001239053.1 | 14 | 3872 | ||
GenBank mRNA | AL117400.1 | - | 6 | 4172 | ||
BC050565.1 | - | 15 | 3831 | 4537404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
