siRNA Details
Gene Symbol: B330016D10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046111.1 | - | 1 | 1505 | ||
AK046532.1 | - | 1 | 1502 | |||
AK080800.1 | - | 1 | 1513 | |||
AK080864.1 | - | 1 | 1482 | |||
AK082239.1 | - | 1 | 884 | |||
AK148126.1 | - | 1 | 854 |
