siRNA Details
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122952.1 | NP_001116424.1 | 11 | 8330 | ||
NM_001122953.1 | NP_001116425.1 | 10 | 8440 | |||
NM_010905.3 | NP_035035.1 | 11 | 8569 | |||
GenBank mRNA | AK034669.1 | - | 1 | 1938 | ||
AK043885.1 | - | 1 | 1690 | |||
AK051150.1 | - | 1 | 1217 | |||
AK053150.1 | - | 1 | 1638 | |||
AK173173.1 | - | 9 | 5255 | |||
BC094331.1 | - | 1 | 1340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
