siRNA Details
Gene Symbol: Smg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031814.1 | NP_001026984.1 | 2 | 575 | ||
GenBank mRNA | AK041264.1 | - | 2 | 354 | ||
AK047829.1 | - | 2 | 556 | |||
AK049391.1 | - | 2 | 575 | |||
AK157798.1 | - | 2 | 562 | |||
AK220328.1 | - | 2 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
