siRNA Details
Gene Symbol: CLDN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124076.1 | - | 2 | 1098 | ||
AK126315.1 | - | 2 | 3562 | |||
BC000671.2 | - | 1 | 1130 | 3349211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
