siRNA Details
Gene Symbol: Sycp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177191.3 | NP_796165.2 | 34 | 3463 | ||
GenBank mRNA | AK029735.1 | - | 7 | 774 | ||
AK036624.1 | - | 12 | 1311 | |||
DQ103262.1 | - | 34 | 3463 |

More Information
Other Species Targeted

Gene Ontology Categories:
