siRNA Details
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045545.1 | - | 1 | 1221 | ||
AK078989.1 | - | 1 | 1080 | |||
AK143106.1 | - | 2 | 209 | |||
BC027113.1 | - | 2 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
