siRNA Details
Gene Symbol: Zfp629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177226.5 | NP_796200.1 | 3 | 649 | ||
GenBank mRNA | AK031161.1 | - | 3 | 358 | ||
AK034484.1 | - | 3 | 321 | |||
AK172933.1 | - | 4 | 379 | |||
BC035547.1 | - | 3 | 291 | 4481313 | ||
BC067053.1 | - | 3 | 649 | 30543652 |
