siRNA Details
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133758.3 | NP_598519.2 | 3 | 508 | ||
NM_177249.3 | NP_796223.2 | 4 | 568 | |||
GenBank mRNA | AK041541.1 | - | 3 | 488 | ||
AK082114.1 | - | 4 | 532 | |||
AK083763.1 | - | 4 | 785 | |||
AK151051.1 | - | 3 | 512 | |||
BC108425.1 | - | 4 | 509 | 30062566 | ||
BY743916.1 | - | 3 | 508 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107542, XM_006230044, XM_006230046, XM_006230045, BC166609.
Gene Ontology Categories:
