siRNA Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271463.1 | NP_001258392.1 | 21 | 2688 | ||
NM_001271464.1 | NP_001258393.1 | 20 | 2607 | |||
NM_001271465.1 | NP_001258394.1 | 21 | 2715 | |||
NM_177266.5 | NP_796240.3 | 21 | 2721 | |||
GenBank mRNA | AK035819.1 | - | 21 | 2626 | ||
AK044371.1 | - | 20 | 2608 | |||
AK083441.1 | - | 20 | 2727 | |||
BC057062.1 | - | 10 | 2499 | |||
BC138233.1 | - | 21 | 2617 | 8861252 | ||
BC145208.1 | - | 21 | 2619 | 9053759 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
