siRNA Details
Gene Symbol: CSTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326.2 | NP_001317.1 | 4, 5 | 423 | ||
GenBank mRNA | AK290836.1 | - | 4, 5 | 423 | ||
AK294204.1 | - | 1, 2 | 16 | |||
AK297371.1 | - | 3, 4 | 305 | |||
BC108319.1 | - | 4, 5 | 397 | 6197136 | ||
U15782.1 | - | 4, 5 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
